Learn More
Organomation 80 Position MULTIVAP Nitrogen Evaporator, 220 volt wiring
SDP
Supplier:  Organomation 118802
Encompass_Preferred
Request Bulk Order
The 80 position MULTIVAP parallel evaporator is a high temperature dry block model. This instrument contains a rectangular gas distribution system mounted on a frame and dual post assembly. The manifold delivers nitrogen gas to the needles or glass pipettes, which direct the gas onto the sample’s surface. This method results in rapid solvent evaporation and sample concentration. In order to conserve nitrogen gas, the manifold has a toggle switch for every row of 8 samples, which allows individual rows to be shut off when evaporating batches of less than 80 samples. Nitrogen gas flow is also controlled by an adjustable flow meter and a built-in pressure regulator.
Catalog No. NC1983170