Learn More
Organomation 8 Position ROT-X-TRACT-LC Extractor
SDP
Supplier:  Organomation 13318
Encompass_Preferred
Request Bulk Order
The ROT-X-TRACT-LC is designed to accommodate up to eight (8) Corning Accelerated One-Step apparatuses. The Corning Accelerated One-Step extraction apparatus combines continuous liquid-liquid extraction and concentration into one step. A modified Friedrichs condenser is used to reduce overall height to accommodate traditional laboratory hood specifications; however, Allihn condensers can be substituted per customer request. The hydrophobic membrane allows the organic solvent to circulate through the 1-liter sample bottle continuously until the extraction is complete. By incorporating a stop-cock assembly and 3-ball Snyder column, the Corning One-Step extractor can both extract desired analytes as well as evaporate solvent from the end by-product within the same cycle. The Corning One-Step extractor comes with an electronic side control box which enables the operator to program specific bath temperatures and time cycles to stay in compliance with the designated methodologies.
Catalog No. NC2301764