Learn More
Ace Glass, Inc. Double tube vacuum manifold, 730mm long, (4) stopcock ports with 24/40 inner joints
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 873842
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)