Learn More
Ace Glass, Inc. 6000mL jacketed pressure reactor, complete with #15 Ace-Thred inlet/outlets, 150mm Ace-Thred 6 neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 6386528
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
6000mL jacketed pressure reactor, complete with #15 Ace-Thred inlet/outlets, 150mm Ace-Thred 6 neck head, 80mm bottom valved adapter, 100 micron PE & 350 micron PP filter supports, stir shaft, bearing, condenser, addition funnel, clamp & O-rings