Learn More
Chemglass Life Sciences 600mL Freeze Dry Flask, Virtis Style, P/N 149088
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG161124
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
600mL Freeze Dry Flask, Virtis Style, P/N 149088. Flat bottom borosilicate flask for use with and Virtis freeze dryers. Wide mouth design provides easy access sample.