Learn More
Chemglass Life Sciences 6in X 6in X 1/8in Thick Quartz Plate
Supplier: Chemglass Life Sciences CGQ062020
6in X 6in X 1/8in Thick Quartz Plate. Commercial quality ground and polished clear fused quartz discs. Edges have a ground finished on all sizes. SIZES OTHER THAN THOSE LISTED ARE AVAILABLE. PLEASE CALL FOR INFORMATION.