Learn More
Chemglass Life Sciences 4.65" ID x 6.64" OD x 1.20" Height White PTFE Support Ring
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG305506
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
4.65" ID x 6.64" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING