Learn More
JDM SCIENTIFIC LLC 5UL CHECKIT GO
Supplier: JDM SCIENTIFIC LLC MSLO8V56
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The LabBee Checkit Go is a simple, stand-alone, disposable cartridge that allows you to check the accuracy of your liquid handling robot or multi-channel pipette in just seconds. No additional equipment is requiredThe easy to use LabBee branded Checkit Go device measures the dispensed volumes of up to 8 channels simultaneously in less than 10 seconds. The cartridge is the size of a well plate, and can be used in your automated liquid handler or on a lab bench.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.