Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Bromo-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™
$79.00 - $343.00
Specifications
CAS | 162607-17-2 |
---|---|
Melting Point | 135°C |
Molecular Formula | C4H4BBrO2S |
InChI Key | USJPOBDLWVCPGG-UHFFFAOYSA-N |
SMILES | B(C1=CC=C(S1)Br)(O)O |
Specifications
162607-17-2 | |
C4H4BBrO2S | |
B(C1=CC=C(S1)Br)(O)O | |
206.848 | |
206.85 |
135°C | |
USJPOBDLWVCPGG-UHFFFAOYSA-N | |
(5-bromothiophen-2-yl)boronic acid | |
2734319 | |
Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title