Learn More
Medchemexpress LLC HY-10586 200mg Medchemexpress, 5-Azacytidine CAS:320-67-2 Purity:>98%
Supplier: Medchemexpress LLC HY10586200MG
Medchemexpress, HY-10586 200mg 5-Azacytidine CAS:320-67-2 5-Azacytidine (Azacitidine; 5-AzaC; Ladakamycin) is a nucleoside analogue of cytidine that specifically inhibits DNA methylation. 5-Azacytidine is incorporated into DNA to covalently trap DNA methyltransferases and contributes to reverse epigenetic changes[2]. 5-Azacytidine induces cell autophagy[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.