Learn More
Genesis Bps LLC 4″Blood Shoe, 20 per pack
Supplier: Genesis Bps LLC 4003544
Positions blood bags upright. Promotes proper air circulation. Clear design enables data capture without removing units of blood. Acrylic material is durable, long lasting and can be easily cleaned.