Learn More
Electron Microscopy Sciences Multi Holder 45 degree for 6 pin stubs, Hitachi M4
Supplier: Electron Microscopy Sciences 7595270H
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This 35 mm in diameter 45 Degree multi pin holder is designed to save time. It allows for a higher SE signal without having to tilt the specimen. It is made from machined aluminum with stainless steel Allen set screws. It accommodates up to six standard 12.7 mm (1/2") pin stubs.
Type: Hitachi M4