Learn More
R S Crum & Company 42G SERIES BALL VALVE
Supplier: R S Crum & Company SS42GVCR4
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
STAINLESS STEEL 42G SERIES BALL VALVE, 1/4" MALE VCR ENDS, PTFE SEALS, CV=0.60
This item is not returnable.
View return policy