Learn More
Ace Glass, Inc. Vacuum manifold, 405mm long, (4) stopcock ports with 14/20inner joints
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 874018
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)