Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water and DMSO.
Notes
Store at 4°C. Incompatible with oxidizing agents. Avoid excessive heat and light.
4-Amino-6-hydroxy-2-mercaptopyrimidine is a novel, potent, and selective A3 adenosine receptor antagonists. It is also used as pharmaceutical intermediates.
Solubility
Soluble in water and DMSO.
Notes
Store at 4°C. Incompatible with oxidizing agents. Avoid excessive heat and light.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Quantity | 100 g |
Beilstein | 5795441 |
Solubility Information | Soluble in water and DMSO. |
Formula Weight | 161.19 (143.17 Anhydrous) |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate |
Safety and Handling
EINECSNumber | 213-722-8 |
RTECSNumber | YQ8817000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction