Learn More
Ace Glass, Inc. Pycnometer Tube, 4.5mL, Specific gravity
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 543712
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Provides a method for measurement of light hydrocarbon liquids as per ASTM D 941. Also Ind. and Eng. Chemistry, Analytical edition, Vol. 16, No. 1, Jan. 1944, page 55.