Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Versaflo™ TR-600 Series Filter Covers
For use with Versaflo™ TR-600 Series PAPR systems.
Supplier: 3M™ 7100021251
This item is not returnable.
View return policy
Description
- Filter cover helps protect cartridge from physical damage
- Easy to use cover enables use of prefilters and/or spark arrestors
- Allows PAPR to be worn through decontamination showers or rain
- Also used to hold TR-6600 Prefilter or TR-662 Spark Arrestor in place
- Color: Gray
- 3M Catalog# TR-6500FC , 3M Catalog# TR-6700FC
Applications:
Recommended Industries: Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing
Certifications
NIOSH Approved
Compliance
NIOSH approved.
Specifications
NIOSH | |
Filter Cover | |
TR-6700 and TR-600 Respirators, TR-6600 Prefilter, and TR-662 Spark Arrestor | |
1/Cs. |
Gray | |
Healthcare, Pharmaceutical, Heavy Industrial, Manufacturing, General Manufacturing | |
TR-6700FC |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction