Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3B Scientific™ Kundts Tube
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Demonstrate standing sound waves using this glass tube and calculate wavelengths of sound using a method devised by Kundt.
Supplier: 3B Scientific™ U8432845
Includes:
- One Glass tube with funnel
- One plunger
- One filler chute
- One whistle
- One bottle of cork powder
Description
- Cork powder is used calculate wavelength of sound
- Effective length of the tube can be altered by means of a piston
- Length: 600mm
- External diameter: 20mm
- Internal diameter: 17mm
Specifications
Kundts Tube | |
One Glass tube with funnel, one plunger, one filler chute, one whistle, one bottle of cork powder | |
60 cm | |
0.35 kg |
For demonstrating standing sound waves and calculating wavelengths of sound using cork powder in a method devised by Kundt | |
23.6 in. | |
0.77 lb. | |
1700 Hz |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction