Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3-[(tert-Butoxycarbonyl)amino]phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
$61.00 - $212.00
Specifications
CAS | 380430-68-2 |
---|---|
Melting Point | 168°C |
Molecular Formula | C11H16BNO4 |
MDL Number | MFCD03411945 |
InChI Key | CWLNHPXWZRALFS-UHFFFAOYSA-N |
Specifications
380430-68-2 | |
C11H16BNO4 | |
CWLNHPXWZRALFS-UHFFFAOYSA-N | |
[3-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic acid | |
2773228 | |
Crystalline Powder |
168°C | |
MFCD03411945 | |
B(C1=CC(=CC=C1)NC(=O)OC(C)(C)C)(O)O | |
237.062 | |
237.06 |
Safety and Handling
TSCA : No
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title