Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Cyanopyrazolo[1,5-a]pyrimidine, 97%, Thermo Scientific™

Catalog No. AAH5838004
Click to view available options
Quantity:
2 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25939-87-1
Molecular Formula C7H4N4
Molecular Weight (g/mol) 144.137
MDL Number MFCD01927799
InChI Key RRHORVAOECWFPT-UHFFFAOYSA-N
Synonym pyrazolo 1,5-a pyrimidine-3-carbonitrile, 3-cyanopyrazolo 1,5-a pyrimidine, 8-hydropyrazolo 1,5-a pyrimidine-3-carbonitrile, pyrazolo 1,5-a-pyrimidine-3-carbonitrile, 2vtm, pyrazolo 1,5-a pyrimidin-3-yl cyanide, pyrazolo 1,5-a pyrimidine-3-carbonitrile, 8, lzm
PubChem CID 819275
IUPAC Name pyrazolo[1,5-a]pyrimidine-3-carbonitrile
SMILES C1=CN2C(=C(C=N2)C#N)N=C1

Specifications

Quantity 2 g
UN Number UN3439
Formula Weight 144.14
Percent Purity 97%
Chemical Name or Material 3-Cyanopyrazolo[1,5-a]pyrimidine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.