Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Chloro-4-(4-morpholinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific™

Catalog Number AAH59229MD
Click to view available options
Quantity:
250 mg
1361110-63-5
C16H23BClNO3
323.62
MFCD18070979
SKJAWVMUHVUHMM-UHFFFAOYSA-N
4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-chloro-4-morpholinophenylboronic acid pinacol ester, 3-chloro-4-morpholinophenylboronicacidpinacolester, 3-chloro-4-4-morpholinyl benzeneboronic acid pinacol ester, 3-chloro-4-morpholinophenyl boronic acid pinacol ester, 4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl morpholine, 4-2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine
56965741
4-[2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(C=C1)N1CCOCC1
1361110-63-5
C16H23BClNO3
323.62
MFCD18070979
SKJAWVMUHVUHMM-UHFFFAOYSA-N
4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine, 3-chloro-4-morpholinophenylboronic acid pinacol ester, 3-chloro-4-morpholinophenylboronicacidpinacolester, 3-chloro-4-4-morpholinyl benzeneboronic acid pinacol ester, 3-chloro-4-morpholinophenyl boronic acid pinacol ester, 4-2-chloro-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl phenyl morpholine, 4-2-chloro-4-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl morpholine
56965741
4-[2-chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]morpholine
CC1(C)OB(OC1(C)C)C1=CC(Cl)=C(C=C1)N1CCOCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 323.63
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-(4-morpholinyl)benzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit