Learn More
Roboz Surgical 3-0 SILK BLACK BRAIDED
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Roboz Surgical SUT153
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Suture Material,Non-Sterile, 100Yard Spool. Size: 3-0; Silk, Black, Braided, Non-Absorbable