Learn More
Jensen Inert Products 2L ALTEF Gas Sampling Bag with ATT MP + Flap
Supplier: Jensen Inert Products GSSP1S0909MPE
ALTEF is a proprietary fluoropolymer film developed especially for the Gas Sampling market. ALTEF does not exhibit background levels of DMAC or phenol. ALTEF is chemically inert to most acids, aliphatic and aromatic organic compounds, chlorinated solvents, and alcohols. ALTEF has low VOC background and is suitable for most VOC's and many sulfur compounds. Longer sample storage times than most other bag materials. ALTEF is not recommended for ketones or esters in high concentrations (>30%) and is not suitable for storing H2S. Maximum operating temperature: 260 ºF
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.