Learn More
Chemglass Life Sciences 250mL Glassblowers Heavy Wall Round Bottomed Flask Blank
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG61806HW
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
250mL Glassblowers Heavy Wall Round Bottomed Flask Blank. Heavy Wall blanks are 30% heavier than Chemglass's standard CG-618 series. Blanks are used in the fabrication of single or multi-neck round bottom flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.