Learn More
CELLTREAT Scientific Products 250mL Filter System, 0.45m PES Filter, 75mm Diameter, Sterile
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: CELLTREAT Scientific Products 229702
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
CELLTREAT Scientific Products 250mL, 0.45¿m PES Filter System. Easy-to-read molded graduations marks on bottle top filter. Bold easy-to-read printed membrane and pore size for easy identification. Curved bottle design for easy handling. Printed lot number and expiration date on every bottle top. Improved filter support provides faster flow rates. Angled hose barb adapter fits three size hose diameters and prevents the filter from tipping. Polyethersulfone (PES) membrane is for high flow and low binding. 0.45¿m PES membrane ideal for solution clarification. Filter Systems include bottle top filter and solution bottle with cap. Large diameter filter increases flow and prevents clogging. Membrane diameter of 75mm. Every case is printed with the lot number for quality traceability. Packaged sterile, individually bagged, 12/case.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.