Learn More
Across International 250C UL Certified 1.9 CF Vacuum Oven 5 Sided Heat SST Tubing
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Across International AT19UL
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Introducing the new Ai UL certified 1.9 cubic foot vacuum ovens, this certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan.Also come standard is the secondary over-temp protection dial, all stainless steel tubing/compression fittings, stainless steel vacuum/vent valve, oil-filled vacuum gauge, 5 sided pad heating technology and two year warranty. The thermal-conductive aluminum shelves provide excellent temperature uniformity inside the chamber, while our 4th gen LCD low proportional gain temperature controllers keep your oven temperature accurate.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.