Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Thiazolidinedione, 99%, Thermo Scientific Chemicals

Catalog Number AAA1349622
Click to view available options
Quantity:
25 g
100 g
2295-31-0
C3H3NO2S
117.122
MFCD00005478
ZOBPZXTWZATXDG-UHFFFAOYSA-N
2,4-thiazolidinedione, thiazolidinedione, thiazolidine-2,4-dione, 2,4-dioxothiazolidine, 2,4 3h,5h-thiazoledione, usaf ek-5496, thiazolidindione, unii-aa68lxk93c, 2,4 thiazolidinedione, thiazolidinedione-2,4
5437
CHEBI:50992
1,3-thiazolidine-2,4-dione
C1C(=O)NC(=O)S1
2295-31-0
C3H3NO2S
117.122
MFCD00005478
ZOBPZXTWZATXDG-UHFFFAOYSA-N
2,4-thiazolidinedione, thiazolidinedione, thiazolidine-2,4-dione, 2,4-dioxothiazolidine, 2,4 3h,5h-thiazoledione, usaf ek-5496, thiazolidindione, unii-aa68lxk93c, 2,4 thiazolidinedione, thiazolidinedione-2,4
5437
CHEBI:50992
1,3-thiazolidine-2,4-dione
C1C(=O)NC(=O)S1

2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Thiazolidinedione may be used in the synthesis of sixteen 5-arylidene-2,4-thiazolidinediones, via ultrasound-promoted aldol condensation reaction. It may be used as starting material for the synthesis of drugs with antihyperglycemic activity.

Solubility
Soluble in water (30 mg/ml at 3°C), methanol, DMSO, ethanol, and ethyl ether.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 126°C
Boiling Point 178°C to 179°C (19 mmHg)
Quantity 100 g
Beilstein 110700
Solubility Information Soluble in water (30mg/ml at 3°C),methanol,DMSO,ethanol,and ethyl ether.
Formula Weight 117.13
Percent Purity 99%
Chemical Name or Material 2,4-Thiazolidinedione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-941-2
RTECSNumber XJ5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit