Learn More
Chemglass Life Sciences 15L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1965615
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
15L Process Reactor, Complete, Electric Motor, 70.1" OAH with Motor, 15.3" Clearance Below Valve. Features:• Support Frame with Oversized Rugged Stainless Steel Uprights and Heavy Duty PTFE Coated Aluminum Fittings• Improved Support Clamp Auto-Centers Reactor and Lid• Perfluoro O-Rings On 60mm and 200mm Flanges• Stainless Steel Inlet/Outlet Adapters Allow Connection To Cooling/Heating Circulator• Zero Dead Space Detachable Drain Valve Has Knob with Laser Etched Directional Arrows For Open and Close• Electric or Air Motor. Optional XP Motor Available• 5-Neck Lid with Large Powder Addition Funnel• Digital Temperature Monitor• Electric Motor Has Separate Digital Controller• Reactor Jacket Graduated On Both Sides• Removable Stainless Steel Pipe Used To Support Side Shelf and Manifold System• Support Bushings For Optional Safety Shield• Heavy Duty Lockable Casters
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.