Learn More
Organomation Unheated 15 Position MICROVAP Nitrogen Evaporator
Supplier: Organomation 11815O
The unheated 15 position MICROVAP benchtop evaporator is intended for small batch solvent evaporations, such as concentrations of sample groups in microcentrifuge tubes. It offers an effective and compact evaporation solution for samples in life science and pharmaceutical laboratories. This model has no heat for evaporations that will be performed at ambient temperature.Each unheated MICROVAP uses minimal hood space while using Ambient temperature for heat sensitive applications. The 15 position MICROVAP comes standard with stainless steel luer lock hubs and 4-inch x 19-gauge stainless steel needles for easy removal.Up to 15 samples can be accommodated at once, and the instrument comes customized for one sample size with an outside diameter of 10-22mm. If multiple sample sizes will be used with the instrument, inserts can be purchased for each additional sample size. Each of the sample blocks and inserts are customized to your samples for a precise fit.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.