Learn More
Diagnostic Biosystems 10X TRIS-EDTA RETRIEVAL BUFFER 10X Tris-EDTA Buffer For Heat Induced Epitope Recovery, pH 9.0
Supplier: Diagnostic Biosystems K043
10X Tris-EDTA Buffer For Heat Induced Epitope Recovery, pH 9.0 is designed for use during the heat induced epitope retrieval (HIER) step prior to immunohistochemistry on formalin-fixed paraffin embedded tissue sections. The use of this buffer in combination with heat (often by microwave, water bath, or pressure cooker) has been shown to restore the antigenicity of proteins modified during the formalin fixation of tissue. This buffer is supplied as a 10X stock solution.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.