Learn More
Chemglass Life Sciences 10L Jacketed Cylindrical Process Filter Reactor System Complete, Electric Motor
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1959F610
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
10L Jacketed Cylindrical Process Filter Reactor System Complete, Electric Motor. Chemglass's jacketed filter glass reactors can be used for single or multi-step reactions and filtrations in the same vessel, production and process scale peptide synthesis. Filter base eliminates material transfers and significantly reduces losses. Complete reactor system on Chemglass's open air, autocenter stand. Please Note: Filter Reactors are for Vacuum/Gravity Filtration Use Only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.