Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-4-(trifluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. B177225G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 407-14-7
Molecular Formula C7H4BrF3O
Molecular Weight (g/mol) 241.01
MDL Number MFCD00040834
InChI Key SEAOBYFQWJFORM-UHFFFAOYSA-N
Synonym 1-bromo-4-trifluoromethoxy benzene, 4-trifluoromethoxy bromobenzene, 4-bromo trifluoromethoxy benzene, 4-trifluoromethoxybromobenzene, p-bromotrifluoromethoxybenzene, benzene, 1-bromo-4-trifluoromethoxy, 4-bromo-1-trifluoromethoxy benzene, p-trifluoromethoxybromobenzene, 4-bromotrifluoromethoxybenzene, 4-bromo-trifluoromethoxybenzene
PubChem CID 521008
IUPAC Name 1-bromo-4-(trifluoromethoxy)benzene
SMILES FC(F)(F)OC1=CC=C(Br)C=C1

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 g
Formula Weight 241.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-(trifluoromethoxy)benzene
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.