Learn More
Glass Expansion PTFE Sheathed Carbon Fibre Probe 0.25mm ID with EzyFit Connector (for Cetac ASX-520/560/280 and PerkinElmer S20 Sereis Autosamplers)
Supplier: Glass Expansion 708031088
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
PTFE Sheathed Carbon Fiber Probe 0.25mm ID with EzyFit Connector. Suitable for Cetac ASX-200/500/800 and PerkinElmer S20 Series Autosamplers. EzyFit connector connects probe capillary directly to Glass Expansion F-Series nebulizers.Replacement of autosampler probe is recommended about every 6 months to a year. This will ensure you have unrestricted sample flow. If clogs within probes or nebulizers occur, consider using the Guardian In-Line Sample Filter (P/N NC0897716, 70-803-1108). The Guardian In-Line Sample Filter is spliced into the autosampler probe capillary and can be backflushed using the Eluo Nebulizer Cleaner for Glass Concentric Nebulizers (P/N NC9462559, 70-ELUO), or the Eluo HF Nebulizer Cleaner for OpalMist or DuraMist (P/N 501907353, 70-ELUO-OPD), and Eluo Adaptor for In-Line Filter (P/N NC0801128, 70-803-1160). The Guardian In-Line Sample Filter is reuseable and can last years.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.